missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POMC Monoclonal Antibody (6Y8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA518042
This item is not returnable.
View return policy
Description
This antibody is specific for the 1-24 amino acid sequence in ACTH and has no cross-reactivity with ACTH 17-39. The antibody has an affinity constant greater than 1011/mol.
POMC Monoclonal antibody specifically detects POMC in Human samples. It is validated for Immunohistochemistry (Paraffin).
Specifications
| POMC | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 10mM sodium azide | |
| P01189 | |
| POMC | |
| proprietary immunogen | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| 6Y8 | |
| Unconjugated | |
| POMC | |
| ACTH; adrenal corticotropic hormone; adrenocorticotropic hormone; adrenocorticotropin; alpha-melanocyte stimulating hormone; alpha-melanocyte-stimulating hormone; alphaMSH; alpha-MSH; BE; beta-endorphin; Beta-LPH; beta-melanocyte-stimulating hormone; beta-MSH; CLIP; Corticotropin; Corticotropin-like intermediary peptide; corticotropin-lipotropin; Gamma-LPH; gamma-MSH; lipotr; Lipotropin beta; lipotropin gamma; LPH; Melanocyte-stimulating hormone alpha; Melanocyte-stimulating hormone beta; Melanotropin alpha; melanotropin beta; Melanotropin gamma; met-enkephalin; MSH; Npp; opiomelanocortin prepropeptide; OTTHUMP00000119991; OTTHUMP00000200964; POC; Pomc; Pomc1; Pomc-1; Pomc2; Potential peptide; Precursor of MSH; pro-ACTH-endorphin; proopiomelanocortin; pro-opiomelanocortin; proopiomelanocortin (adrenocorticotropin/ beta-lipotropin/ alpha-melanocyte stimulating hormone/ beta-melanocyte stimulating hormone/ beta-endorphin); proopiomelanocortin preproprotein; proopiomelanocortin, beta (end | |
| Mouse | |
| Protein A | |
| RUO | |
| 5443 | |
| Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction