missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
POMC Monoclonal antibody specifically detects POMC in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
Specifications
| Antigen | POMC |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 967905 |
| Conjugate | FITC |
| Formulation | PBS |
| Gene Alias | ACTH, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte-stimulating hormone, alpha-MSH, beta-endorphin, beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, corticotropin-like intermediary peptide, corticotropin-lipotropin, gamma-LPH, gamma-MSH, lipotropin beta, lipotropin gamma, LPH, melanotropin alpha, melanotropin beta, melanotropin gamma, met-enkephalin, MSH, NPP, POC, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin preproprotein |
| Host Species | Mouse |
| Immunogen | Human Adrenocorticotropic hormone/ACTH) synthetic peptide, Accession # P01189 |
| Purification Method | Protein A or G purified from hybridoma culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?