missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Pomalidomide 5'-fluoro-6'-piperazine

Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Pomalidomide 5'-fluoro-6'-piperazine is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand with a terminal piperazine for onward chemistry to generate Protein Degraders with rigid linkers. Part of a range of functionalized tool molecules for PROTAC™ Degrader R&D.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 2-(2,6-Dioxo-3-piperidinyl)-5-fluoro-6-(1-piperazinyl)-1 H-isoindole-1,3(2 H)-dione hydrochloride |
| CAS | 2222114-23-8 |
| Quantity | 25 mg |
| Target | Degrader Building Blocks |
| Molecular Formula | C17H17FN4O4.HCl |
| Purity | 0.95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction