missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Pomalidomide 4'-PEG5-azide

Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Pomalidomide 4'-PEG5-azide is a functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand plus a PEG5 linker with terminal azide ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 4-[(17-Azido-3,6,9,12,15-pentaoxaheptadecyl)amino]-2-(2,6-dioxo-3-piperidinyl)-1 H-isoindole-1,3(2 H)-dione |
| CAS | 2624181-61-7 |
| Quantity | 25 mg |
| Target | Degrader Building Blocks |
| Molecular Formula | C25H34N6O9 |
| Purity | 0.95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction