missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POM121 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585161
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (82%), Rat (81%). Positive Control: A549. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
POM121 (POM121 membrane glycoprotein), also known as POM121A (nuclear envelope pore membrane protein POM 121A), NUP121 (nucleoporin Nup121) or pore membrane protein of 121 kDa, is a 1,249 amino acid single-pass membrane protein that exists as three alternatively spiced isoforms and belongs to the POM121 family. Encoded by a gene that maps to human chromosome 7q11.23, POM121 is highly conserved and localizes to nucleus membrane and endoplasmic reticulum membrane. An essential component of the nuclear pore complex (NPC), POM121 is mandatory for nuclear envelope formation and may play a role in biogenesis of the NPC. POM121 associates with the central spoke ring complex and is involved in anchoring components of the pore complex to the pore membrane by way of its F-X-F-G repeat-containing domain. POM121 overexpression induces the formation of cytoplasmic annulate lamellae (AL).
Specifications
| POM121 | |
| Polyclonal | |
| Unconjugated | |
| POM121B | |
| 2610027A18Rik; C80273; KIAA0618; mKIAA0618; Nuclear envelope pore membrane protein POM 121; nuclear envelope pore membrane protein POM 121A; nuclear envelope pore membrane protein POM 121C; nuclear pore membrane glycoprotein 121 kD; nuclear pore membrane protein 121; nuclear pore membrane protein 121 kDa; nuclear pore membrane protein 121-2; nucleoporin Nup121; Nup121; p145; Pom121; POM121 membrane glycoprotein; POM121 membrane glycoprotein C; POM121 transmembrane nucleoporin; POM121 transmembrane nucleoporin C; POM121-2; POM121A; POM121B; POM121C; Pore membrane protein of 121 kDa; pore membrane protein of 121 kDa C | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100101267, 100288540, 107939, 9883 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 0.16 mg/mL | |
| PBS with 20% glycerol, 1% BSA and 0.025% ProClin 300; pH 7 | |
| A6NF01, A8CG34, Q8K3Z9, Q96HA1 | |
| POM121, POM121B, POM121C | |
| Recombinant protein encompassing a sequence within the N-terminus region of human POM121. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction