Learn More
BRAND™ POM Joint Clip
Product Code. 10243441 Shop All Brand Products
Click to view available options
Color:
Red
Unit Size:
Each
Product Code. 10243441
Brand: BRAND™ 55646
Only null left
Add to basket
Add to basket
Will not scratch glass surfaces. Excellent resilience and stability.
  • POM construction
  • Outstanding chemical resistance
  • Operating temperature up to 150°C
  • Fits NS 29/32
TRUSTED_SUSTAINABILITY
Specifications
Type Joint Clip
Material POM
Size 29/32
Color Red