Learn More
Cytiva POM/EPDM Packing Connectors
Product Code. 10392730 Shop All Cytiva Products
Click to view available options
For Use With (Equipment):
For use with XK 16 columns
For use with XK 26 columns
Unit Size:
Each
Product Code. 10392730
Brand: Cytiva 18115344
Only null left
Add to basket
Add to basket
Column connectors for use in gel filtraion chromatography applications
This packaging connector used to connect another column, being used as a packing reservoir, to a XK 16 or XK 26 column to achieve efficient packing. A properly packed media bed is essential for obtaining optimal results in all forms of liquid chromatography, particularly in gel filtration. The packing connector makes it easy to obtain a well-packed bed of gel filtration and other media in XK 16 and XK 26 Columns, and is especially convenient for fast and easy packing of affinity, IEX, and HIC media to short bed heights.
TRUSTED_SUSTAINABILITY
Specifications
Type Packing Connector
Temperature (Metric) Max 60°C
Includes 2 x Sealing ring are delivered with the packing connector
Material EPDM, POM
For Use With (Equipment) For use with XK 16 columns