missing translation for 'onlineSavingsMsg'
Learn More

Polyvinylsulfuric acid potassium salt, approx. M.W. 175,000

Product Code. 10025980 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10025980 25 g Glass bottle 25g
10702032 5 g Glass bottle 5g
11976971 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10025980 Supplier Thermo Scientific Acros Supplier No. 418140250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26837-42-3 | (C2H3KO4S)n | NaN g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26837-42-3
Molecular Formula (C2H3KO4S)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00081923
InChI Key WZAPMUSQALINQD-UHFFFAOYSA-M
Synonym pvsk, potassium ethenyl sulfate, potassium poly vinyl sulfate, polyvinyl sulfate potassium salt, sulfuric acid, monovinyl ester, polymers, potassium salt, sulfuric acid, monoethenyl ester, homopolymer, potassium salt, vinyl sulfate potassium salt, sulfuric acid vinyl=potassium salt
PubChem CID 5055241
SMILES [K]OS(=O)(=O)OC(-*)C-*

Specifications

Color Beige-Brown to White
Physical Form Crystalline Powder
Linear Formula (-CH2CH(OSO3K)-)n
Quantity 25 g
IUPAC Name potassium ethenyl sulfate
Percent Purity ≥21.2% (K)
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Polyvinylsulfuric acid potassium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.