missing translation for 'onlineSavingsMsg'
Learn More

Polyvinylpyrrolidone, MedChemExpress

Product Code. 16453806
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16453806 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16453806 Supplier Medchem Express Supplier No. HYB1620
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Polyvinylpyrrolidone is a compound which has been widely tested and used in human and veterinary medicine as an effective wound healing accelerator and disinfectant when combined with iodine and other compounds.

Polyvinylpyrrolidone is a compound which has been widely tested and used in human and veterinary medicine as an effective wound healing accelerator and disinfectant when combined with iodine and other compounds.

Specifications

Chemical Name or Material Polyvinylpyrrolidone
CAS 9003-39-8
Color White
Purity Grade Notes Research
Molecular Formula (C6H9NO)n
Quantity 25 g
Synonym PVP Polyvidone Povidone
Solubility Information H2O : ≥ 50 mg/mL ∣DMSO : 50 mg/mL (Need ultrasonic)
SMILES O=C1CCCN1C(C[H])[H].[n]
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.