missing translation for 'onlineSavingsMsg'
Learn More

Polyvinylpyrrolidone K30, MP Biomedicals™

Product Code. 11406162 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Unit Size:
100g
1kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11406162 100 g 100g
11416162 500 g 500g
11426162 1 kg 1kg
11436162 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11406162 Supplier MP Biomedicals Supplier No. 02102786.1
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Used as a thickener and as a dispersing agent. Polyvinylpyrrolidone K30, MP Biomedicals is a component of Denhardt's Solution. It is also used as a detoxicant, a complexing agent, a lubricant and a binder.

Chemical Identifiers

CAS 9003-39-8
Molecular Formula (C6H9NO)n
Molecular Weight (g/mol) 111.14
MDL Number MFCD01076626
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
PubChem CID 6917
ChEBI CHEBI:82551
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES *-CC(-*)N1CCCC1=O

Specifications

Density 1.23–1.29g/mL (Lit.)
pH 2.8 to 4.2 (5% aq. soln.)
Quantity 100 g
Solubility Information Soluble in water (>100mg/mL, 2.5% aq solution), methanol, ethanol, alcohol, chloroform and glycerol, acetic acid; insoluble in dimethyl ether, ethyl acetate, acetone, toluene, xylene, mineral oil, carbon tetrachloride.
Viscosity 27–35cps (1% aq soln)
Formula Weight 35,000–51,000
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.