missing translation for 'onlineSavingsMsg'
Learn More

Polyvinylpyrrolidone, Fisher BioReagents™

Product Code. 10102873
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
Product Code. Quantity Packaging unitSize
10102873 500 g Poly Bottle 500g
10619173 100 g Poly Bottle 100g
2 options
This item is not returnable. View return policy

Product Code. 10102873

Brand: Fisher BioReagents BP431500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Polyvinylpyrrolidone, >-95%, C6H9NO, CAS Number-9003-39-8, n-vinyl-2-pyrrolidinone, 2-pyrrolidinone, 1-ethenyl, n-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, 1-vinylpyrrolidone, 1-vinyl-2-pyrrolidinone, 1-vinyl-2-pyrrolidone, n-vinylpyrrolidone, n-vinylpyrrolidinone, vinylpyrrolidone, 500g

Polyvinylpyrrolidone, >-95%, C6H9NO, CAS Number-9003-39-8, n-vinyl-2-pyrrolidinone, 2-pyrrolidinone, 1-ethenyl, n-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, 1-vinylpyrrolidone, 1-vinyl-2-pyrrolidinone, 1-vinyl-2-pyrrolidone, n-vinylpyrrolidone, n-vinylpyrrolidinone, vinylpyrrolidone, 500g

Polyvinyl Pyrrolidone is a component of Denhardt's Reagent, commonly used in nucleic acid hybridization procedures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-39-8
Molecular Formula (C6H9NO)n
Molecular Weight (g/mol) 111.14
MDL Number MFCD01076626
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
PubChem CID 6917
ChEBI CHEBI:82551
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES *-CC(-*)N1CCCC1=O

Specifications

Color White
pH 3 to 7
Physical Form Solid
Melting Point 100°C
Ash 0.05% max.
Assay Percent Range ≥95 %
Merck Index 15, 7814
Quantity 500 g
Percent Purity ≥95%
Identification Pass Test
Packaging Poly Bottle
Assay (dry basis)≥95%
Chemical Name or Material Polyvinylpyrrolidone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.