missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyvinylpyrrolidone, average M.W. 8.000, K16-18, Thermo Scientific Chemicals
39.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White or White-Yellow |
pH | 3.0 to 5.0 (5% aq. soln.) |
Physical Form | Powder or Flakes |
Melting Point | 130.0°C |
Merck Index | 15, 7814 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform, practically insoluble in ether |
Quantity | 500 g |
Flash Point | >215°C |
Percent Purity | 11.5 to 12.8% (nitrogen, on anh. subst.), 10ppm 1-vinylpyrrolidin-2-one max. (HPLC) |
Identification | Positive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'