missing translation for 'onlineSavingsMsg'
Learn More

Polyvinylpyrrolidone, average M.W. 8.000, K16-18, Thermo Scientific Chemicals

Catalog Number 10114333
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
9003-39-8
(C6H9NO)n
111.14
MFCD01076626
WHNWPMSKXPGLAX-UHFFFAOYSA-N
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
6917
CHEBI:82551
1-ethenylpyrrolidin-2-one
*-CC(-*)N1CCCC1=O
9003-39-8
(C6H9NO)n
111.14
MFCD01076626
WHNWPMSKXPGLAX-UHFFFAOYSA-N
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
6917
CHEBI:82551
1-ethenylpyrrolidin-2-one
*-CC(-*)N1CCCC1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White or White-Yellow
pH 3.0 to 5.0 (5% aq. soln.)
Physical Form Powder or Flakes
Melting Point 130.0°C
Merck Index 15, 7814
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform, practically insoluble in ether
Quantity 2.5 kg
Flash Point >215°C
Percent Purity 11.5 to 12.8% (nitrogen, on anh. subst.), 10ppm 1-vinylpyrrolidin-2-one max. (HPLC)
Identification Positive
Infrared Spectrum Authentic
Packaging Plastic bottle
Water 5% Max. (K.F.)
Chemical Name or Material Polyvinylpyrrolidone, K16-18
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'