missing translation for 'onlineSavingsMsg'
Learn More

Polyvinylpyrrolidone, average M.W. 1.300.000, K85-95

Product Code. 10115163 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
Packaging:
Plastic Drum
Plastic bottle
Unit Size:
1kg
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10115163 2.5 kg Plastic Drum 2.5kg
10125163 25 g Plastic bottle 25g
10696822 500 g Plastic bottle 500g
10713041 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10115163 Supplier Thermo Scientific Acros Supplier No. 227470025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9003-39-8 | (C6H9NO)n | 111.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-39-8
Molecular Formula (C6H9NO)n
Molecular Weight (g/mol) 111.14
MDL Number MFCD01076626
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
PubChem CID 6917
ChEBI CHEBI:82551
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES *-CC(-*)N1CCCC1=O

Specifications

Color Cream-White to White
pH 4 to 7 (5% aq. soln.)
Physical Form Powder
Merck Index 15, 7814
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform, practically insoluble in ether
Quantity 2.5 kg
Percent Purity 12 to 12.8% (nitrogen)
Identification positive
Infrared Spectrum Authentic
Packaging Plastic Drum
Water 5% max. (K.F.)
Chemical Name or Material Polyvinylpyrrolidone, Average M.W. 1.300.000, K85-95
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.