missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Polyvinyl Chloride Stirring Rods

Product Code. p-8007254
Click to view available options
Length (Metric):
250 mm
300 mm
350 mm
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 10460163

Brand: Kartell™ 0049800

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Minimalize scratching with a rod constructed of PVC for durability. Kartell™ Polyvinyl Chloride Stirring Rods are the perfect substitue for a glass stirring rod.

  • Rigid
  • Lightweight
  • Unbreakable
  • Ideal replacement for glass rods

Specifications

Color White
Diameter (Metric) 7.15 mm
Length (Metric) 300 mm
Material PVC
Quantity 10/Pk.
Type Stirring Rod
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.