Learn More
Cytiva Polyvinyl Chloride Fume Cupboard
This rich, robust, reliable, fume cupboard made of polyvinylchloride features glare fluorescent tube lightening.
POA
Specifications
Product Type | Fume Cupboard |
---|---|
Lighting Type | Glare Fluorescent Tube |
Material | PVC |
Quantity | 1 Ea. |
Description
- Fume cupboard made of polyvinylchloride with glare fluorescent tube lightening
- Standard sorbridge PVC (= registry fume cupboards)
- PVC rear panel over the entire width of the fume hood
- Guide the air at the top and bottom
- Uses all types of solvents, volatile products, powders, condensing
Specifications
Fume Cupboard | |
Glare Fluorescent Tube | |
PVC | |
1 Ea. |