missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyvinyl Alcohol, Fully Hydrolyzed, 99.0 to 99.8%, BAKER™, J.T. Baker™
Click to view available options
Quantity:
1 kg
Packaging:
Wide Mouth Amber Glass Bottle
Specifications
CAS | 9002-89-5 |
Molecular Formula | (C2H4O)n |
MDL Number | MFCD00081922 |
Synonym | vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval |
Quantity | 1 kg |
InChI Key | IMROMDMJAWUWLK-UHFFFAOYSA-N |
SMILES | OC(-*)C-* |
IUPAC Name | ethenol |
Molecular Weight (g/mol) | 44.05 |
PubChem CID | 11199 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'