missing translation for 'onlineSavingsMsg'
Learn More

Polyvinyl alcohol, 87-89% hydrolyzed, high molecular weight

Product Code. 11359607
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359607 100 g 100g
11349607 25 g 25g
11369607 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11359607 Supplier Thermo Scientific Alfa Aesar Supplier No. 041240.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol

Polyvinyl alcohol is used as a biological reagent to test the dispersion of zinc oxide nanoparticles. It is an effective protein resistant material and is used to stabilize gold colloids of diameter 3-8nm. It is used as an emulsion polymerization aid to make polyvinyl acetate, as a protective colloid, as a film used in the water transfer printing process, as a surfactant for the formation of polymer encapsulated nanobeads, as an embolization agent in medical procedures and as a fixative for specimen collection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-89-5
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 44.05
MDL Number MFCD00081922
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
Synonym vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
PubChem CID 11199
IUPAC Name ethenol
SMILES OC(-*)C-*

Especificaciones

Physical Form Solid
Solubility Information Soluble in water.
Quantity 100 g
Percent Purity 87 to 89%
Odor Odorless
Chemical Name or Material Polyvinyl alcohol, High molecular weight, Hydrolyzed

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.