Learn More
Honeywell™ POLYTRIL™ Top Gloves
Description
- Knitted polyamide glove
- Black oilproof nitrile coating on palm and back
- Fully dipped
- Smooth finish
- Elastic wrist
- The knitted construction conforms perfectly to the natural shape of the hand thus providing excellent dexterity
- Open back to provide superior ventilation
- Polyamide combines good resistance to abrasion, humidity, tearing and shearing
- The nitrile coating on the palm and fingertips brings some excellent abrasion resistance quality as well as oilproof properties
- A pattern finish to enhance grip
- Black color dedicated to dirty applications and environments
Specifications
Specifications
| Certifications/Compliance | EN 420-2003 Basic standards, EN 388-2016 Mechanical hazards |
| Description | 13 gauge |
| Coating Material | Polyamide, Nitrile |
| Color | Black |
| Size | 9 |
| Length (Metric) | 25 cm |