Learn More
Honeywell™ POLYTRIL™ Air 3/4 Gloves
Product Code. 15896715
Click to view available options
Size:
7
8
9
Unit Size:
1 pair
Product Code. 15896715
Brand: Honeywell™ 223227209
Only null left
Add to basket
Add to basket
Honeywell™ POLYTRIL™ Air 3/4 Gloves for handling of fine products in a greasy, damp or dirty atmosphere.
  • Black polyamide knitted glove
  • Black foam nitrile coating on the palm and fingers
  • Open back
  • Elasticated knitted wrist
  • The ergonomics seamless knit construction conforms better to the natural shape of the hand for greater strength and durability, superior confort and ventilation
  • The filament yarn reduce linting for the most sensitive applications and easily laundered for extended use
  • The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • The foamy coating and ventilated back optimise the glove's breathability
  • The nitrile foam improves the grip
  • The knitted and elasticated wrist gives the glove good support
  • The nitrile coating at the palm and fingertips provides outstanding abrasion resistance and impermeability to oils
  • The black color is suitable for work in a soiled atmosphere
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE CATEGORY : II Intermediary risks In accordance with standards: EN 420-2003 Basic standards EN 388-2016 Mechanical hazards
Description 13 gauge
Coating Material Nitrile Foam
Color Black
Size 9
Length (Metric) 24.5 cm