Learn More
Honeywell™ POLYTRIL™ Air 3/4 Gloves
Click to view available options
Size:
7
8
9
Unit Size:
1 pair
Description
- Black polyamide knitted glove
- Black foam nitrile coating on the palm and fingers
- Open back
- Elasticated knitted wrist
- The ergonomics seamless knit construction conforms better to the natural shape of the hand for greater strength and durability, superior confort and ventilation
- The filament yarn reduce linting for the most sensitive applications and easily laundered for extended use
- The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
- The foamy coating and ventilated back optimise the glove's breathability
- The nitrile foam improves the grip
- The knitted and elasticated wrist gives the glove good support
- The nitrile coating at the palm and fingertips provides outstanding abrasion resistance and impermeability to oils
- The black color is suitable for work in a soiled atmosphere
Specifications
Specifications
| Certifications/Compliance | CE CATEGORY : II Intermediary risks In accordance with standards: EN 420-2003 Basic standards EN 388-2016 Mechanical hazards |
| Description | 13 gauge |
| Coating Material | Nitrile Foam |
| Color | Black |
| Size | 7 |
| Length (Metric) | 24.5 cm |