missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Shermond™ Polythene Gloves
Click to view available options
Size:
Small
Medium
Unit Size:
Pack of 100
Description
- Inexpensive disposable gloves provide adequate covering for light work
- Ambidextrous
- Unpowdered
- Clear polythene
- Not sterile
- Not suitable for use with chemicals
Specifications
Specifications
| Certifications/Compliance | None |
| Color | Clear |
| Exterior Finish | No Texture |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Material | Polyethylene |
| Powdered | Powder Free |
| Product Type | Gloves |
| Size | Medium |
| Sterility | Non-sterile |
| Show More |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction