missing translation for 'onlineSavingsMsg'
Learn More

Polytetrafluoroethylene, 6-10 micron, Thermo Scientific Chemicals

Product Code. 11339678
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339678 1 kg 1kg
11349678 50 g 50g
11329678 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11339678 Supplier Thermo Scientific Alfa Aesar Supplier No. 044184.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Polytetrafluoroethylene, 6-10 micron is used in specially designed coating applications on coil for bakeware.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-84-0
Molecular Formula (C2F4)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00084455
SMILES FC(F)(-*)C(F)(F)-*

Specifications

Physical Form Powder
Merck Index 14,7583
Solubility Information Insoluble in water. Soluble in perfluoro kerosene.
Quantity 1 kg
Particle Size 6 to 10 Micron
Chemical Name or Material Polytetrafluoroethylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.