missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polysulfide Oil Standard: 15,000 μg/g S in Mineral Oil, VHG™
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Description
Polysulfide Oil Standard: 15,000 μg/g S in Mineral Oil
Specifications
Specifications
| Methods | XRF |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | 13 cSt Mineral Oil |
| Quantity | 1 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction