Learn More
Polystyrene Test Film
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 1 Ea. |
| Chemical Name or Material | Polystyrene Test Film |
Frequently Asked Questions (FAQs)
The test film is 19 mm in diameter and 38 microns thick. The holder is 5.08 cm (2 inches) x 10.2 cm (4 inches).
Polystyrene Test Film is used to calibrate IR machines.