missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Technical Service Consultants™ Polystyrene Swabs with Viscose Tip in Tube
Click to view available options
Quantity:
500Pack
Unit Size:
Pack of 500
Description
- Extensive range of swabs available with a choice of swab shafts including white birchwood, polypropylene (flexible) and polystyrene (breakable)
- Tips are formed from pure cotton, alginate and cotton
- Gamma irradiated to ensure a SAL of at least 10-6
Specifications
Specifications
| Includes | Shaft with Viscose Tip In Tube |
| Certifications/Compliance | CE Marked: Class IIa |
| Shelf Life | 2 Years |
| Tip Material | Polystyrene |
| Packaging | Tube |
| For Use With (Application) | General Sampling, useful for All Organisms |
| Quantity | 500Pack |
| Product Type | Swab |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction