Learn More
Greiner Bio-One Polystyrene Standard Lids for Microplates
Product Code. 10292811
Change view
Click to view available options
Plate Compatibility:
384-well Standard Microplate
96-well Microplate, Non TC-treated
High-format Low-profile Microplates, 384-well LoBase Small Volume or 1536-well Microplate
Sterility:
Non-sterile
Sterile
Format:
High profile
Low profile
Ultra low profile
Unit Size:
Pack of 100
Pack of 200
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plate Compatibility Sterility Format Unit Size
10292811 384-well Standard Microplate Sterile Low profile Pack of 200
10389031 384-well Standard Microplate Non-sterile Low profile Pack of 200
10421611 96-well Microplate, Non TC-treated Non-sterile High profile Pack of 100
10414625 High-format Low-profile Microplates, 384-well LoBase Small Volume or 1536-well Microplate Sterile Ultra low profile Pack of 100
10761211 96-well Microplate, Non TC-treated Sterile High profile Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10292811 Supplier Greiner Bio-One Supplier No. 656191
Estimated Shipment: 19-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Provides protection against contamination and evaporation during sample storage and cell cultivation. Greiner BioOne™ Polystyrene Standard Lids are available in a wide variety of formats to meet laboratory needs.
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • High lid profile; height: 9 mm
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Polystyrene
Sterility Sterile
Includes Chimney-well style 96 well plates
Condensation Rings No
No. per Pack 200
Format Low profile
Product Type Plate Lid
Plate Compatibility 384-well Standard Microplate