Learn More
Lonza™ Polystyrene Sample Container, 15 mL Tubes
For transporting product samples for endotoxin analysis or sample storage.
Brand: Lonza™ BE2099
Code : NEW
Additional Details : Weight : 0.02000kg
Description
- Intended for transporting product samples for endotoxin analysis or sample storage.
- Proper container and storage conditions need to be validated for each individual sample.
- These sample containers offer a greater capacity at a reduced cost.
- Polystyrene construction.
Specifications
Sample Container | |
Polystyrene | |
15 mL tubes | |
50/Pk. |