Learn More
Lonza™ Polystyrene Sample Container, 15 mL Tubes
Product Code. 11595759
Click to view available options
No. per Pack:
50/Pk.
Unit Size:
Pack of 50
Product Code. 11595759
Brand: Lonza™ BE2099
Currently Unavailable: We have no information when this product will be available.
View alternative products
For transporting product samples for endotoxin analysis or sample storage.
  • Intended for transporting product samples for endotoxin analysis or sample storage.
  • Proper container and storage conditions need to be validated for each individual sample.
  • These sample containers offer a greater capacity at a reduced cost.
  • Polystyrene construction.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Sample Container
Material Polystyrene
For Use With (Equipment) 15 mL tubes
No. per Pack 50/Pk.