missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Polystyrene Petri Dishes

Product Code. 11719452
Change view
Click to view available options
Dimensions (Dia. x H):
35 x 10 mm
60 x 15 mm
90 x 15 mm
94 x 15 mm
94 x 16 mm
100 x 20 mm
120 x 17 mm
145 x 20 mm
Sterility:
Non-sterile
Sterile
Vent:
No
Yes
Quantity:
120 Pack
240 Pk.
360 Pack
420 Pack
480 Pack
560 Pack
600 Pack
740 Pack
Unit Size:
Pack of 120
Pack of 240
Pack of 360
Pack of 420
Pack of 480
Pack of 560
Pack of 600
Pack of 740
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (Dia. x H) Sterility Vent Quantity unitSize
11719452 120 x 17 mm Sterile No 240 Pk. Pack of 240
10470613 94 x 16 mm Non-sterile No 480 Pack Pack of 480
10639021 90 x 15 mm Non-sterile Yes 420 Pack Pack of 420
10083251 100 x 20 mm Non-sterile Yes 360 Pack Pack of 360
10548071 120 x 17 mm Non-sterile Yes 240 Pk. Pack of 240
10048770 35 x 10 mm Non-sterile Yes 740 Pack Pack of 740
11779442 60 x 15 mm Non-sterile Yes 560 Pack Pack of 560
10340111 145 x 20 mm Non-sterile Yes 120 Pack Pack of 120
10398981 94 x 15 mm Non-sterile Yes 480 Pack Pack of 480
10364451 94 x 16 mm Non-sterile Yes 480 Pack Pack of 480
10669781 94 x 15 mm Non-sterile Yes 480 Pack Pack of 480
10707801 60 x 15 mm Non-sterile Yes 600 Pack Pack of 600
11739452 94 x 15 mm Sterile Yes 480 Pack Pack of 480
10787112 94 x 16 mm Sterile Yes 480 Pack Pack of 480
10742312 94 x 16 mm Sterile Yes 480 Pack Pack of 480
10668631 94 x 15 mm Non-sterile Yes 480 Pack Pack of 480
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Product Code. 11719452 Supplier Greiner Bio-One Supplier No. 688161
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured from highly transparent polystyrene for its heat resistant properties to enable use with hot agar. Greiner BioOne™ Polystyrene Petri Dishes are available in a wide variety of formats and sizes to accommodate multiple research needs.

  • Heat resistant up to 60°C
  • Available in vented and non-vented
  • Choose from standard round design, square-profile dish, special models and contact dishes/OneWell Plate™ models
  • Easy stacking
  • Crystal polystyrene for optimal analysis by microscopy
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Disposable Single-use
Material Polystyrene
Shape Square
Sterility Sterile
Quantity 240 Pk.
Vent No
Dimensions (Dia. x H) 120 x 17 mm
Height (Metric) 17 mm
Diameter (Metric) 120 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.