missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Polystyrene Vented Petri Dishes
GREENER_CHOICE

Product Code. 10204092
Change view
Click to view available options
Sterility:
Sterile
Diameter (Metric):
35 mm
60 mm
145 mm
Height (Metric):
10 mm
15 mm
20 mm
Unit Size:
Pack of 120
Pack of 600
Pack of 740
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Diameter (Metric) Height (Metric) unitSize
10204092 Sterile 35 mm 10 mm Pack of 740
10043341 Sterile 145 mm 20 mm Pack of 120
10459381 Sterile 60 mm 15 mm Pack of 600
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10204092 Supplier Greiner Bio-One Supplier No. 627161
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Used for easy stacking. Greiner Bio-One Polystyrene Vented Petri Dishes is designed for improved gas exchange.

  • Vented for improved gas exchange
  • Manufactured from crystal polystyrene for optimal analysis by microscopy
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Culture Area 8.5 cm2
Material Polystyrene
Shape Round
Sterility Sterile
Type Petri Dish
No. per Case 740
Vent Yes
Dimensions (Dia. x H) 35 x 10 mm
Height (Metric) 10 mm
Diameter (Metric) 35 mm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.