missing translation for 'onlineSavingsMsg'
Learn More

Polystyrene latex microsphere, 1 micron, dry form

Product Code. 15476545
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15476545 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15476545 Supplier Thermo Scientific Alfa Aesar Supplier No. 043711.ME
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol

Polystyrene is used in adhesive, plastics, coatings, for electrical applications. In laboratories containers such as tubes, petridishes were made in polystyrene, used for the building construction panel packing, insulators, for injection molding and casting, as sheets, plates, rods, rigid forms and beads..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 9003-53-6
Physical Form Microsphere
Molecular Formula (C8H8)n
MDL Number MFCD00084450
Synonym ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
Solubility Information Soluble in benzene,toluene,pyridine,dichloromethane,tetra hydro furan,methyl isobutyl ketone. Insoluble in water.
Quantity 500 mg
InChI Key PPBRXRYQALVLMV-UHFFFAOYSA-N
SMILES *-CC(-*)C1=CC=CC=C1
IUPAC Name styrene
Molecular Weight (g/mol) 104.15
PubChem CID 7501
ChEBI CHEBI:27452
Odor Odorless
Particle Size 1 Micron
Chemical Name or Material Polystyrene latex microsphere, Dry form
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.