missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polystyrene latex microsphere, 0.10 micron, 2.5 wt% dispersion in water, Thermo Scientific Chemicals
Click to view available options
Quantity:
15 mL
Description
Polystyrene latex microsphere is covalently attached to a single-stranded DNA in order to increase the charge stabilization of the microspheres. Further, it is used as an efficient support for immunological reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS | 9003-53-6 |
Physical Form | Dispersion |
Molecular Formula | (C8H8)n |
MDL Number | MFCD00084450 |
Synonym | ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene |
Solubility Information | Soluble in methylethylketone,benzene and toluene. Slightly soluble in diethyl ether,n-octanol and acetone. Insoluble in water. |
Quantity | 15 mL |
InChI Key | PPBRXRYQALVLMV-UHFFFAOYSA-N |
SMILES | *-CC(-*)C1=CC=CC=C1 |
IUPAC Name | styrene |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'