missing translation for 'onlineSavingsMsg'
Learn More

Polystyrene latex microsphere, 0.05 micron, 2.5 wt% dispersion in water, Thermo Scientific Chemicals

Product Code. 15476505
Click to view available options
Quantity:
10 mL

 

Polystyrene latex microsphere, 0.05 micron, 2.5 wt% dispersion in water is used in protein binding and as size markers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Color White
CAS 9003-53-6
Physical Form Dispersion
Molecular Formula (C8H8)n
MDL Number MFCD00084450
Synonym ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
Solubility Information Soluble in methylethylketone,benzene and toluene. Slightly soluble in diethyl ether,n-octanol and acetone. Insoluble in water.
Quantity 10 mL
InChI Key PPBRXRYQALVLMV-UHFFFAOYSA-N
SMILES *-CC(-*)C1=CC=CC=C1
IUPAC Name ethenylbenzene
Molecular Weight (g/mol) 104.15
PubChem CID 7501
ChEBI CHEBI:27452
Concentration or Composition (by Analyte or Components) 2.5 wt% dispersion in water
Odor Odorless
Particle Size 0.05 Micron
Chemical Name or Material Polystyrene latex microsphere
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'