Learn More
Macherey-Nagel™ Polystyrene Divinylbenzene Chromabond LV-HR-X
Brand:  Macherey-Nagel™ 732130
Code : 50
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 12880961
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
30 mg
45 or 85 μm
1 to 14
30 Pack
Chromabond LV-HR-X
Polystyrene Divinylbenzene
55 - 60 Å