missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Polystyrene Cell Culture Dish with Vent
Brand: Greiner Bio-One 664920
Additional Details : Weight : 0.80000kg
Specifications
Clear | |
Polystyrene | |
100 mL | |
58 sq. cm | |
Round | |
Fibronectin | |
16 to 17 mL | |
Lid | |
100 mm |
Closure Included | |
Flat | |
Clear | |
Polystyrene | |
Non-sterile | |
Cell Culture Dish | |
Increase of isolation and cultivation efficiency, low-serum or serum-free cultivation, cell adhesion studies, Promotion of cell adhesion, proliferation and growth of endothelial cells, fibroblasts, smooth muscle cells, neurons and epithelial cells | |
20 mm |