missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Polystyrene 384-Well Microplate
GREENER_CHOICE

Product Code. 10194083 Shop All Corning Products
Change view
Click to view available options
Well Shape:
Flat Bottom (F-well)
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Well Shape unitSize
10194083 Flat Bottom (F-well) Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10194083 Supplier Corning™ Supplier No. 3643
  • ONLINE EXCLUSIVE PRICE
    £3565.00
    £3368.80 / Pack of 50
    Save £196.20
    5% Off
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Suited for fluorescent and luminescent assays using either top or bottom detection microplate readers.

Suited for fluorescent and luminescent assays using either top or bottom detection microplate readers.

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Culture Area 0.03 cm2
Array 16 x 24
Color Black
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 5 to 40 μL
Sterility Sterile
Total Volume (Metric) Well 50 μL
Well Shape Flat Bottom (F-well)
For Use With (Application) For miniaturized 384 well fluorescent cell-based assays
No. per Pack 50Pack
Quantity 50/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.