Learn More
Greiner Bio-One μClear™ 384-well, Cell Culture-Treated, Flat-Bottom Microplate
GREENER_CHOICE
Product Code. 10180422
Change view
Click to view available options
Packaging:
20/Pk., 120/Cs.
8/Pk.
Unit Size:
Pack of 120
Pack of 32
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging unitSize
10180422 8/Pk. Pack of 32
10013393 20/Pk., 120/Cs. Pack of 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10180422 Supplier Greiner Bio-One Supplier No. 781091
Only null left
Add to basket
Add to basket
Color: Black, Clear Bottom; Sterility: Sterile; Lid: With Lid
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic, non-cytotoxic
  • Compatible with automated systems
  • Stackable
  • Alphanumeric well coding μClear™ bottom
  • Well profile: F-bottom (flat bottom)
  • Growth area per well: 10 mm²
  • Working volume per well: 15 μl - 110 μl
  • Available with  lid that enables gas exchange with the lowest possible evaporation
  • Sterile
TRUSTED_SUSTAINABILITY
Specifications
Barcode Alphanumeric
Format Standard
Lid With Lid
Array 16 x 24
Packaging 8/Pk.
Color Black, Clear Bottom
Material Polystyrene
No. of Wells 384
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Rounded Square
Quantity 8/Pk.