missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 80

Product Code. 11378026
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11378026 2500 mL 2.5 L
11358026 100 mL 100mL
11368026 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11378026 Supplier Thermo Scientific Alfa Aesar Supplier No. L13315.0F
  • ONLINE EXCLUSIVE PRICE
    £157.00
    £141.65 / 2.5 L
    Save £15.35
    9% Off
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9005-65-6 | (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | 604.82 g/mol

Used for solubilizing membrane proteins during isolation and purification.Polysorbate 80 is used as a surfactant in soaps, cosmetics, body washes, hair shampoos, hair loss treatments, lotions, creams, bath oils and in mouthwash in the health care industry. It serves as an emulsifier in foods such as ice cream and in the preparation of antiarrhythmic amiodarone. Further, it acts as a solubilizing agent and excipient in the pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-65-6
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
Molecular Weight (g/mol) 604.82
MDL Number MFCD00082107
InChI Key HDTIFOGXOGLRCB-KTKRTIGZNA-N
Synonym Polyoxyethylene sorbitan monooleate
IUPAC Name 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Boiling Point >100°C
Density 1.064
Merck Index 14,7582
Solubility Information Miscible with water,ethanol,cottonseed oil,corn oil,ethyl acetate,methanol and toluene. Immiscible with mineral and vegetable oils.
Quantity 2500 mL
Flash Point >149°C (300°F)
Refractive Index 1.475
Chemical Name or Material Polysorbate 80

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.