missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 80 (Technical Grade), TRC

Product Code. 30012571
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012571 25 g 25g
30011033 5 g 5g
30011591 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012571 Supplier Toronto Research Chemicals Supplier No. TRCP68943325G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-65-6
Molecular Formula C32H60O10
Molecular Weight (g/mol) 604.8
Synonym Polyethoxylated Sorbitan Monooleate, Polyethylene Glycol Sorbitan Ether Monooleate, Polyethylene Glycol Sorbitan Monooleate, Polyoxyethylated Sorbitan Monooleate, Polyoxyethylene Monosorbitan Monooleate, Polyoxyethylene Sorbitan Monooleate, Polyoxyethylenesorbitan Oleate, Tween 80
SMILES [HO(CH2CH2O)w]C1COC([CH(OCH2CH2)yOH]CO[(CH2CH2O)z-1])[C@H]1[(OCH2CH2)OH].O=C(CCCCCCC/C=C/CCCCCCCC)OC[CH2]

Specifications

Quantity 25 g
InChI Formula InChI=1S/C32H60O10/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-30(36)40-25-24-37-27-29(39-22-19-34)32-28(38-21-18-33)26-31(42-32)41-23-20-35/h9-10,28-29,31-35H,2-8,11-27H2,1H3/b10-9+
Chemical Name or Material Polysorbate 80
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.