missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 40

Product Code. p-7046570
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 11398116

Brand: Thermo Scientific Alfa Aesar L15030.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9005-66-7 | (C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6 | 522.68 g/mol

Polysorbate 40 has been used in a study to assess the determination of iodide in urine based on a chemiluminescence system. It has also been used in a study to investigate the delivery of astaxanthin and canthaxanthin to HepG2 cells. Also used as oil-in-water emulsifiers in pharmaceuticals, cosmetics, cleaning compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-66-7
Molecular Formula (C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165345
InChI Key DVRFSZFBSOKHQW-UHFFFAOYNA-N
Synonym tween r 40, unii-4nr71fn0ia, unii-sti11b5a2x, 4nr71fn0ia, sti11b5a2x, sorbitan, monohexadecanoate, poly oxy-1,2-ethanediyl derivs., polysorbate 40 usan:inn:ban:nf, sorbitan, monopalmitate polyoxyethylene deriv.
PubChem CID 92329579
IUPAC Name 2-[(2R)-2-[(2R,3R,4R)-3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Density 1.09
Solubility Information Fully miscible with water. Soluble in isopropyl alcohol (100mg/ml),and xylene. Insoluble in propylene glycol,and mineral oil.
Quantity 100 mL
Refractive Index 1.47
Chemical Name or Material Polysorbate 40

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.