missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ratiolab™ Yellow Pipet Tip E
Description
These pipette tips offer uniform high quality and precision. Together with specially developed Multirack, Refill System and the ratiolab™ Multibox with the associated pipet tips in Multitray, make pipetting much simpler and safe in the most sensitive applications.
- Made of high quality polypropylene
- Tight fit due to the precisely fitting cone
- Optimal handling due to the stable shape
- Autoclavable
- Guaranteed dust-free due to special ionization treatment
- Available pre-sterilized or non-sterile
- Excellent transparency due to high quality surfaces
- Minimal residual wetting due to controlled quality of raw materials
- Excellent volume accuracy due to precise tip orifice
- Practice-oriented tip volumes
- Aeroject™ tips with special filters made of hydrophobic PE-HD as
- Aerosol barrier and for protection against contamination
- Suitable for all commonly used pipet types
Specifications
Specifications
| Volume (Metric) | 1 to 200 μL |
| Color | Yellow |
| Autoclavable | Autoclavable |
| Length (Metric) | 4.55cm |
| Certifications/Compliance | CE, IVD |
| For Use With (Application) | Ideal for All Commercially Available Pippets |
| Sterilization Method | Autoclavable |
| For Use With (Equipment) | Universal Fit |
| Material | Polypropylene |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?