missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heinz Herenz Polypropylene Waste Bags BIOHAZARD Red
Click to view available options
Capacity (Metric):
22 L
Unit Size:
Each
Description
- Waste bags BIOHAZARD red
- For easy identification, for the disposal of contaminated labware, rectangle white indicator patch field turns into dark when autoclaved
- With BIOHAZARD symbol and safety imprint in English, German, French, and Spanish languages
- Autoclavable at 134 °C for 20 minutes (the bag must not be completely closed), thickness 50 μm (0.05 mm)
- PP
Specifications
Specifications
| Capacity (Metric) | 22 L |
| Capacity (English) | 4.84 gal. |
| Color | Red |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| Product Type | Biohazard Waste Bag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction