missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azpack™ Polypropylene Wadless Screw Caps
Click to view available options
Color:
Natural
White
Neck Finish:
R3/20
R3/22
R3/24
R3/28
R3/33
R3/38
R3/48
R3/51
R3/58
R3/63
R3/83
R6/31
Unit Size:
Pack of 50
Description
- For medical usages
- White polypropylene
- Softer texture due to white pigmentation
Specifications
Specifications
| Color | White |
| Material | Polypropylene |
| Quantity | 50/PK |
| Thread Size (Metric) | TF mm |
| Neck Finish | R3/28 |
| Product Type | Wadless Screw Cap |
| For Use With (Equipment) | For BTF-425, 430, 440, 600, 610, 625, 630 series bottles |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction