Learn More
BRAND™ Polypropylene Valve Cap
For closing valve block. Suitable for autoclaving.
Brand: BRAND™ 704554
Code : 03
Additional Details : Weight : 0.01000kg
Description
- Polypropylene construction
- Autoclavable
- For use with seripettor™
Specifications
Valve Cap | |
Polypropylene | |
Seripettor™ / Seripettor™ pro |
Autoclavable | |
25 mL | |
1 Ea. |