Learn More
BRAND™ Polypropylene Valve Cap
For closing valve block. Suitable for autoclaving.
Brand:  BRAND™ 704552
Code : 03
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 10039800
  • £25.10 / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Polypropylene construction
  • Autoclavable
  • For use with seripettor™
Specifications
Specifications
Valve Cap
Polypropylene
Seripettor™ / Seripettor™ pro
Autoclavable
2, 10 mL