Learn More
Kartell™ Polypropylene Universal Tip
Reduce sample retention with a tip that offers high clarity and excellent water-repellent properties. Kartell™ Polypropylene Universal Tip uses precise cerntering to guarantee reliable directionality of the sample when delivered.
Brand:  Kartell™ 0964100
Product Code. 10226721
  • £68.45 / Pack of 960
Description
Reduce sample retention with a tip that offers high clarity and excellent water-repellent properties. Kartell™ Polypropylene Universal Tip uses precise cerntering to guarantee reliable directionality of the sample when delivered.
  • Manufactured from virgin cadmium-free PP
  • High clarity and excellent water-repellent properties designed to keep sample retention within the tip to a minimum
  • The precise centering of the orifice at the end of the tip guarantees reliable directionality of the sample when delivered
  • These tips are recommended for use with the Kartell™ automatic micropipettes but deliver excellent results even if used with the most other micropipette brands available on the market
  • Disposable Kartell tips come in different package types to satisfy a variety of needs as economically as possible
Specifications
Specifications
2 to 200 μL
Autoclavable
Rack
Rack
Kartell™, Gilson™, Brand™, Nichiryo™, Biohit™, Socorex™, Eppendorf™, Hamilton™, Gilson™ Pipetman™, Socorex™ Pipettor, Nichiryo™, Brand™ Pipettor, Sartorius™ Biohit™ Pipettor, Eppendorf™ Pipettor, Hamilton™ Pipettor
Yellow
CE/IVD
Universal
960/Pack
Polypropylene