Learn More
Kartell™ Plastilab™ Polypropylene Universal Test Tube Racks
Special design allows samples to be incubated at same temperature.
Brand:  Kartell™ 0056806
Product Code. 15855537
  • ONLINE EXCLUSIVE PRICE
    £288.00
    £219.13 / Pack of 20
    Save £68.87
    23% Off
Only null left
Estimated Shipment: 27-01-2026
Add to basket
Description
Description
Special design allows samples to be incubated at same temperature.
  • Autoclavable at 121°C
  • Large capacity
  • Can be used dry or wet in waterbaths
  • Does not rot, rust, or deform
  • Alphanumerical grid reference on top tier for easy identification
  • Color-coded for bath work