missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Polypropylene Tissue Grinder

Product Code. 10313385 Shop All SP Scienceware Products
Click to view available options
Unit Size:
Pack of 6
This item is not returnable. View return policy

Product Code. 10313385

Brand: Bel-Art™ SP Scienceware™ F199210001

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Polypropylene tissue grinder efficiently homogenizes tissue and cells in microcentrifuge, centrifuge and grinding tubes.

  • Cone tip tapers from 8.2mm (0.33 in.) down to 2.9mm (0.12 in.) in a length of 19.8mm (0.78 in.) and has a rounded end
  • Shaft diameter: 4.7mm (³⁄16 in.)
  • Steam autoclavable at 121°C (250°F)

Specifications

Item Description Polypropylene Tissue Grinder
Autoclavable Autoclavable
Color White
For Use With (Equipment) Microcentrifuge, centrifuge and grinding tubes
Length (Metric) 21cm
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.