Learn More
Masterflex™ Fitting, Polypropylene, TEE-Connector, Hose Barb Union
Product Code. 15770528
Click to view available options
Material:
Polypropylene
Unit Size:
Pack of 10
Product Code. 15770528
Brand: Masterflex™ 3061351
Only null left
Add to basket
Add to basket
Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 3/16 in.
Specifications
Description TEE-Connector, polypropylene, L/S 15-25
Autoclavable Autoclavable
Material Polypropylene
Packaging Quantity 10/pk.
Outlet Port 3
Port Size 3-ports each 3.16 in.
Sterilization Method Autoclave, Ethylene Oxide